Merck
NLRP10 Affects the Stability of Abin-1 To Control Inflammatory Responses.
Journal of immunology (Baltimore, Md. : 1950) (2018-12-05)
Nora Mirza, Anna S Sowa, Katja Lautz, Thomas A Kufer
ABSTRAKT
NOD-like receptors (NLR) are critical regulators of innate immune signaling. The NLR family consists of 22 human proteins with a conserved structure containing a central oligomerization NACHT domain, an N-terminal interaction domain, and a variable number of C-terminal leucine-rich repeats. Most NLR proteins function as cytosolic pattern recognition receptors with activation of downstream inflammasome signaling, NF-κB, or MAPK activation. Although NLRP10 is the only NLR protein lacking the leucine rich repeats, it has been implicated in multiple immune pathways, including the regulation of inflammatory responses toward Leishmania major and Shigella flexneri infection. In this study, we identify Abin-1, a negative regulator of NF-κB, as an interaction partner of NLRP10 that binds to the NACHT domain of NLRP10. Using S. flexneri as an infection model in human epithelial cells, our work reveals a novel function of NLRP10 in destabilizing Abin-1, resulting in enhanced proinflammatory signaling. Our data give insight into the molecular mechanism underlying the function of NLRP10 in innate immune responses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution