Merck
PD-L1 expression in pancreatic adenosquamous carcinoma: PD-L1 expression is limited to the squamous component.
Pathology, research and practice (2018-11-28)
Masahiko Tanigawa, Yoshiki Naito, Jun Akiba, Akihiko Kawahara, Yoshinobu Okabe, Yusuke Ishida, Hiroto Ishikawa, Toru Hisaka, Fumihiko Fujita, Masafumi Yasunaga, Takahiro Shigaki, Tomoya Sudo, Yutaro Mihara, Masamichi Nakayama, Reiichiro Kondo, Hironori Kusano, Kazuhide Shimamatsu, Koji Okuda, Yoshito Akagi, Hirohisa Yano
ABSTRAKT
We examined the programmed death-ligand 1 (PD-L1) expression in surgically resected pancreatic adenosquamous carcinoma (PASC) samples. Furthermore, the detection rate was also assessed using biopsy cases obtained from endoscopic ultrasound-guided fine needle aspiration (EUS-FNA). Fifteen cases of PASC (six resected and nine EUS-FNA biopsied) from the Kurume University Hospital between 2009 and 2016 were used for the evaluation of PD-L1 expression. As a control group, 34 cases of pancreatic ductal adenocarcinomas (PDACs) were selected. To compare the positivity and intensity of PD-L1, two types of clones (SP263, E1L3N) were examined for immunostaining. Only the membrane expression of PD-L1 was regarded as positive. The PD-L1 expressions in the squamous cell carcinoma component (SCc), adenocarcinoma component (ACc), and immune cells were assessed separately. The ratio of PD-L1 expression was calculated by counting the positive tumor cells, and tumor proportion score (TPS) was applied (TPS; Null < 1%, low expression; 1 ≤ TPS ≤ 49% and high expression; ≥ 50%). PD-L1 expression was observed in five surgical PASC samples (83%). This shows that SCc presented a high expression in these cases. However, the overall TPS indicated a low expression. In contrast, only one case (3%) was positive for PD-L1 in PDACs, and the TPS indicated a low expression. No differences in PD-L1 expression were observed between the two clones, SP263 and E1L3N. High PD-L1 expression in the EUS-FNA sample was found in only one case (11%). Although assessment using the tumor cells of PASC samples obtained from EUS-FNA was difficult, this study suggests the selective expression of PD-L1 in the SCc of PASC. Furthermore, it was considered that immune checkpoint inhibitors could provide therapeutic effects selectively on the SCc for the entire range of TPSs, though the PD-L1 expression was low.