Merck
Affinity of Electrochemically Deposited Sol⁻Gel Silica Films towards Catecholamine Neurotransmitters.
Sensors (Basel, Switzerland) (2019-02-23)
María Porcel-Valenzuela, Francisco Huerta, Emilia Morallón, Francisco Montilla
ABSTRAKT
Dopamine, norepinephrine, and epinephrine neurotransmitters can be detected by electrochemical oxidation in conventional electrodes. However, their similar chemical structure and electrochemical behavior makes a difficult selective analysis. In the present work, glassy carbon electrodes have been modified with silica layers, which were prepared by electroassisted deposition of sol⁻gel precursors. These layers were morphologically and compositionally characterized using different techniques, such as field emission scanning electron microscopy (FESEM), TEM, FTIR, or thermogravimetric analysis⁻mass spectrometry (TG-MS). The affinity of silica for neurotransmitters was evaluated, exclusively, by means of electrochemical methods. It was demonstrated that silica adsorbs dopamine, norepinephrine, and epinephrine, showing different interaction with silica pores. The adsorption process is dominated by a hydrogen bond between silanol groups located at the silica surface and the amine groups of neurotransmitters. Because of the different interaction with neurotransmitters, electrodes modified with silica films could be used in electrochemical sensors for the selective detection of such molecules.