Merck
Glypican-1 Promotes Tumorigenesis by Regulating the PTEN/Akt/β-Catenin Signaling Pathway in Esophageal Squamous Cell Carcinoma.
Digestive diseases and sciences (2019-02-08)
Jing Li, Yanjie Chen, Cheng Zhan, Jimin Zhu, Shuqiang Weng, Ling Dong, Taotao Liu, Xizhong Shen
ABSTRAKT
Glypican-1 (GPC1), a cell-surface heparan sulfate proteoglycan, promotes the pathogenesis of many human cancers. This study focuses on the role of GPC1 in the promotion of cell proliferation and motility in esophageal squamous cell carcinoma (ESCC). The expression and distribution of GPC1 were measured in tumor tissues from 248 ESCC patients using immunohistochemical (IHC) assays. Cell counting (kit-8), flow cytometry, Transwell, wound healing, IHC, and Western blotting assays were performed to examine the molecular mechanisms that underlie how GPC1 enhances cell proliferation and motility. The level of GPC1 was higher in ESCC tumor samples than in para-tumor tissues (IHC score: 5.42 ± 2.15 vs. 0.86 ± 0.96). Ectopic overexpression of GPC1 in EC9706 cells promoted cell growth and the G1/S phase transition; conversely, GPC1 knockdown in Eca109 cells attenuated cell proliferation and induced G2/M phase arrest. In addition, GPC1 upregulation enhanced ESCC cell motility and induced epithelial mesenchymal transition (EMT), as demonstrated by the aberrant expression of EMT markers. Mechanistically, we demonstrated that GPC1 increased levels of p-Akt and β-catenin and reduced PTEN expression in ESCC. Our study indicated that GPC1 promotes the aggressive proliferation of ESCC cells by regulating the PTEN/Akt/β-catenin pathway. GPC1 may be a promising target for ESCC treatment.