Merck
  • Human Keratinocytes Use Sphingosine 1-Phosphate and its Receptors to Communicate Staphylococcus aureus Invasion and Activate Host Defense.
Human Keratinocytes Use Sphingosine 1-Phosphate and its Receptors to Communicate Staphylococcus aureus Invasion and Activate Host Defense.
The Journal of investigative dermatology (2019-02-27)
Satomi Igawa, Jae Eun Choi, Zhenping Wang, Yu-Ling Chang, Chia-Chi Wu, Tyler Werbel, Akemi Ishida-Yamamoto, Anna Di Nardo
ABSTRAKT
Sphingosine 1-phosphate (S1P) is a bioactive lipid mediator generated when a cell membrane or its components are damaged by various factors. S1P regulates diverse cell activities via S1P receptors (S1PRs). Keratinocytes express S1PR1-5. Although it is known that S1PRs control keratinocyte differentiation, apoptosis, and wound healing, S1PR functions in keratinocyte infections have not been fully elucidated. We propose that the S1P-S1PR axis in keratinocytes works as a biosensor for bacterial invasion. Indeed, in human impetigo infection, we found high epidermal expression of S1PR1 and S1PR2 in the skin. Furthermore, in normal human epidermal keratinocytes in vitro, treatment with Staphylococcus aureus bacterial supernatant not only induced S1P production but also increased the transcription of S1PR2, confirming our in vivo observation, as well as increased the levels of TNFA, IL36G, IL6, and IL8 mRNAs. However, direct treatment of normal human epidermal keratinocytes with S1P increased the expressions of IL36G, TNFA, and IL8, but not IL6. In both S1P- and S. aureus bacterial supernatant-treated normal human epidermal keratinocytes, S1PR1 knockdown reduced IL36G, TNFA, and IL8 transcription, and the S1PR2 antagonist JTE013 blocked the secretion of these cytokines. Overall, we have proven that during infections, keratinocytes communicate damage by using S1P release and tight control of S1PR1 and 2.