Merck
Systemic Dysfunction of Osteoblast Differentiation in Adipose-Derived Stem Cells from Patients with Multiple Myeloma.
Cells (2019-05-15)
Véronique Béréziat, Christelle Mazurier, Martine Auclair, Nathalie Ferrand, Séverine Jolly, Tiffany Marie, Ladan Kobari, Indira Toillon, François Delhommeau, Bruno Fève, Annette K Larsen, Michèle Sabbah, Laurent Garderet
ABSTRAKT
Keywords: multiple myeloma; bone disease; osteogenesis; adipogenesis; adipose-derived stem cells; bone marrow; senescence; Dickkopf-related protein 1; systemic disease.