Merck
A New Type of Capping Agent in Nanoscience: Metal Cations.
Small (Weinheim an der Bergstrasse, Germany) (2019-04-05)
Jiawei Liu, Shaoyan Wang, Kai Cai, Yefei Li, Zhipan Liu, Lingmei Liu, Yu Han, Hong Wang, Heyou Han, Hongyu Chen
ABSTRAKT
Capping agents are the essential factor in nanoscience and nanotechnology. However, the types of capping agents are greatly limited. Defying conventional beliefs, here is shown that metal cations can also be considered as capping agents for oxide nanoparticles, particularly in maintaining their colloidal stability and controlling their facets. Here the general stabilizing effects of multivalent cations for oxide nanoparticles, and the facet controlling role of Al3+ ions in the growth and ripening of Cu2 O octahedra, are demonstrated. This discovery broadens the view of capping agent and opens doors for nanosynthesis, surface treatment, and beyond.