Merck
Human Indoor Exposome of Chemicals in Dust and Risk Prioritization Using EPA's ToxCast Database.
Environmental science & technology (2019-05-14)
Ting Dong, Yingdan Zhang, Shenglan Jia, Hongtao Shang, Wenjuan Fang, Da Chen, Mingliang Fang
ABSTRAKT
Humans spend most of their time indoors and thus have long-term exposure to chemicals. Dust is a sink for most indoor chemicals, and its ingestion is an important pathway for chemical uptake. Therefore, the chemical atlas from dust is an ideal environmental sample to investigate the indoor exposome and associated risk. In this study, we aimed to establish an indoor exposome database through comprehensive data mining on the occurrence of identified compounds in dust, and we prioritize chemicals of health concern. Through an extensive literature review (2849 articles), 355 chemicals and their concentrations were documented and analyzed for human exposure. Together with 81 compounds without concentration and 75 volatile organic compounds, we have established an indoor exposome database with 511 chemicals. Sixteen toxicological end points were selected for toxicity prioritization. Toxic equivalency factor (TEF)-based toxicity, calculated from EPA's ToxCast database, revealed a comprehensive atlas of the chemicals that had a primary contribution. Many of the prioritized compounds are currently neglected or are not actively studied. Overall, this investigation provides one of the most comprehensive analyses on chemical occurrence in indoor dust and prioritizes their chemical toxicity. Our findings can be used as a database for future exposome studies of the indoor environment and provide guidance for indoor risk assessments.