Merck
Knockdown of serpin peptidase inhibitor clade C member 1 inhibits the growth of nasopharyngeal carcinoma cells.
Molecular medicine reports (2019-03-22)
Jin Xu, Yin Ying, Gaoyun Xiong, Liqin Lai, Qingliang Wang, Yue Yang
ABSTRAKT
Nasopharyngeal carcinoma (NPC) is a type of cancer originating in the nasopharynx. There are no NPC‑specific treatments available at present. Serpin peptidase inhibitor clade C member 1 (SERPINC1) serves roles in anticoagulation and anti‑inflammation. The aim of the present study was to investigate the role of SERPINC1 in the proliferation and apoptosis of NPC cells. Tumor and adjacent healthy tissue samples were collected from patients with NPC. Additionally, the SERPINC1 gene was silenced in the HNE3 cell line using short interfering RNA targeted against SERPINC1 (SERPINC1‑siRNA). Cell viability was determined via a Cell Counting Kit‑8 assay; furthermore, proliferation and apoptosis were investigated via flow cytometry. Western blotting and reverse transcription‑quantitative polymerase chain reaction analysis were performed to determine the expression levels of protein and mRNA. It was revealed that the expression levels of SERPINC1 mRNA and protein were increased in NPC tumor tissues compared with in adjacent healthy tissues. The expression of SERPINC1 mRNA and protein in HNE3 cells decreased following SERPINC1‑siRNA transfection. Furthermore, knockdown of SERPINC1 promoted apoptosis and inhibited proliferation. It was also demonstrated that silencing SERPINC1 upregulated the expression of B‑cell lymphoma-2 (Bcl‑2)‑associated X protein and p53 mRNA and protein, and downregulated that of Bcl‑2, survivin and cyclin D1. Downregulation of SERPINC1 reduced the phosphorylation of phosphatidylinositol 3‑kinase (PI3K), protein kinase B (Akt) and mammalian target of rapamycin (mTOR). Thus, SERPINC1 knockdown may promote the apoptosis of HNE3 cells and inhibit proliferation via the suppression of the PI3K/Akt/mTOR signaling pathway.