Merck
Purification, characterization, antioxidant and moisture-preserving activities of polysaccharides from Rosa rugosa petals.
International journal of biological macromolecules (2018-12-07)
Chaoli Zhang, Fusheng Zhao, Ronghui Li, Yiyan Wu, Shuang Liu, Qichao Liang
ABSTRAKT
The present study, we prepared polysaccharides from Rosa rugosa petals (RRPS) using hot-water extraction and purified the polysaccharides by chromatography to obtain RRPS-1 and RRPS-2. Preliminary structural features of RRPS were characterized by different instrumental methods, such as HPGPC, FT-IR, and GC analysis. The average molecular weights of RRPS-1 and RRPS-2 were 8.8 kDa and 443.8 kDa, respectively. Then, antioxidant and moisture-preserving activities of RRPS were determined in vitro. The analysis of antioxidant activities suggested that RRPS-2 had good potential for scavenging activity of radicals. We also found that the RRPS-2 has strong moisture-preserving activity in vitro. These results clearly indicated that RRPS-2 might have a good potential to be utilized in pharmaceutical, food and cosmetics industries.