Merck
The protective effect of sinapic acid in osteoarthritis: In vitro and in vivo studies.
Journal of cellular and molecular medicine (2019-01-04)
Xiaobin Li, Jian Lin, Xiaoxia Ding, Jiangwei Xuan, Zhichao Hu, Dengying Wu, Xingyu Zhu, Zhenhua Feng, Wenfei Ni, Aimin Wu
ABSTRAKT
The anti-inflammatory effect of sinapic acid (SA) has been reported in several studies. However, whether SA has the same effect on osteoarthritis (OA) has yet to be clearly elucidated. We designed a series of in vitro and in vivo procedures to verify the above conjecture. Compared with controls, SA-pretreated human chondrocytes showed lower levels of interleukin (IL)-1β-induced IL-6, prostaglandin E2 (PGE2), nitric oxide (NO) and tumour necrosis factor-α (TNF-α) in vitro. Meanwhile, SA could also reverse the degradation of type II collage and aggrecan, as well as the overproduction of matrix metalloproteinase-9 (MMP-9) and matrix metalloproteinase-13 (MMP-13), inducible nitric oxide synthase (iNOS), cyclooxygenase (COX)-2 and a disintegrin and metalloproteinase thrombospondin motifs (ADAMTS)-5. Furthermore, activation of nuclear factor κB (NF-κB), which was induced by IL-1β, was also inhibited by SA through the pathway of nuclear factor-erythroid 2-related factor-2 (Nrf2)/heme oxygenase 1. In vivo, SA could delay the progress of mice OA models. We propose that SA may be applied as a potential therapeutic drug in OA treatment.