Merck
  • Combination therapy with polydeoxyribonucleotide and proton pump inhibitor enhances therapeutic effectiveness for gastric ulcer in rats.
Combination therapy with polydeoxyribonucleotide and proton pump inhibitor enhances therapeutic effectiveness for gastric ulcer in rats.
Life sciences (2018-04-18)
Il-Gyu Ko, Sung-Eun Kim, Jun-Jang Jin, Lakkyong Hwang, Eun-Sang Ji, Chang-Ju Kim, Jin-Hee Han, In Taik Hong, Min Seop Kwak, Jin Young Yoon, Hyun Phil Shin, Jung Won Jeon
ABSTRAKT
The main action of proton pump inhibitors (PPIs) is to inhibit gastric acid secretion, and PPIs are widely used to treat gastric ulcer (GU). However, if the action of promoting gastric mucosal regeneration is added, the effectiveness of GU treatment can be enhanced. Thus, in order to improve the therapeutic effect on GU, we tried to develop combination therapy promoting regeneration in injured tissue besides suppressing gastric acid secretion. Polydeoxyribonucleotide (PDRN) was selected to evaluate tissue regeneration, and pantoprazole was chosen as one of the PPIs. GU was induced by oral administration of indomethacin once a day for 7 days. Rats in drug-administered groups were intraperitoneally injected with 100 μL normal saline, containing each drug at the indicated concentration, once a day for 14 days after inducing GU. PDRN and PPI combination therapy potently improved tissue regeneration and inhibited production of pro-inflammatory cytokines. PDRN treatment with or without PPI increased the concentration of cyclic adenosine-3,5'-monophosphate (cAMP) and the ratio of phosphorylated cAMP response element-binding protein (p-CREB) to cAMP response element-binding protein (CREB). PDRN treatment with or without PPI also increased the expressions of vascular endothelial growth factor (VEGF) and adenosine A2A receptor. PDRN and PPI combination therapy showed more potent therapeutic effect on GU compared to the PDRN monotherapy or PPI monotherapy. The excellent therapeutic effect of PDRN and PPI combination therapy on GU appeared by promoting regeneration of damaged tissue as well as inhibiting gastric acid secretion.