- Substrate Elasticity Governs Differentiation of Renal Tubule Cells in Prolonged Culture.
Substrate Elasticity Governs Differentiation of Renal Tubule Cells in Prolonged Culture.
Tissue engineering. Part A (2018-11-30)
Harold D Love, Mingfang Ao, Seiver Jorgensen, Lindsey Swearingen, Nicholas Ferrell, Rachel Evans, Leslie Gewin, Raymond C Harris, Roy Zent, Shuvo Roy, William H Fissell
PMID30484388
ABSTRAKT
Successful clinical tissue engineering requires functional fidelity of the cultured cell to its in vivo counterpart, but this has been elusive in renal tissue engineering. Typically, renal proximal tubule cells in culture have a flattened morphology and do not express key transporters essential to their function. In this article, we show for the first time that in vitro substrate mechanical properties dictate differentiation of cultured renal proximal tubule cells. Remarkably, this effect was only discernable after 4 weeks in culture, longer than usually reported for this cell type. These results demonstrate a new tunable parameter to optimize cell differentiation in renal tissue engineering.