Merck
  • Fabrication of Water-Compatible Molecularly Imprinted Resin in a Hydrophilic Deep Eutectic Solvent for the Determination and Purification of Quinolones in Wastewaters.
Fabrication of Water-Compatible Molecularly Imprinted Resin in a Hydrophilic Deep Eutectic Solvent for the Determination and Purification of Quinolones in Wastewaters.
Polymers (2019-05-16)
Weiyang Tang, Kyung Ho Row
ABSTRAKT
A novel water-compatible molecularly imprinted resin was prepared in a green solvent deep eutectic solvent (DES). Resorcinol and melamine, as functional monomers with an abundant hydrophilic group, such as -OH, -NH2 and -NH-, were introduced into the molecularly imprinted resin (MIR). Three DESs (choline chloride-ethylene glycol, tetramethylammonium bromide-ethylene glycol and tetramethylammonium chloride-ethylene glycol) were used to synthesize the molecularly imprinted resin and the resulting deep eutectic solvent-based molecularly imprinted resins were characterized by particle size analysis, elemental analysis, scanning electron microscopy, Fourier transform infrared spectroscopy and thermogravimetric analysis. The resulting deep eutectic solvent-based molecularly imprinted resins were then applied to the adsorption of quinolones (ofloxacin) in water. The adsorption process of deep eutectic solvent-based molecularly imprinted resin followed the static adsorption model, Langmuir isotherm (R2 ≥ 0.9618) and kinetic model pseudo-second-order (R2 > 0.9814). The highest theory adsorption ability of the three kinds of deep eutectic solvent-based molecularly imprinted resins was more than 23.79 mg/g. The choline chloride-ethylene glycol-based MIR was applied to solid-phase extraction for the determination and purification of quinolones (e.g., ciprofloxacin and ofloxacin). The detection limit of deep eutectic solvent-based molecularly imprinted resin-solid-phase extraction method was less than 0.018 mg/L. The recoveries of the deep eutectic solvent-based molecularly imprinted resin-solid-phase extraction method at three spiked levels were 88.7-94.5%, with a relative standard deviation of ≤4.8%. The novel deep eutectic solvent-based molecularly imprinted resin-solid-phase extraction method is a simple, selective and accurate pre-treatment method and can be used to determine the quinolones in environmental water.