Merck
The NASA Twins Study: A multidimensional analysis of a year-long human spaceflight.
Science (New York, N.Y.) (2019-04-13)
Francine E Garrett-Bakelman, Manjula Darshi, Stefan J Green, Ruben C Gur, Ling Lin, Brandon R Macias, Miles J McKenna, Cem Meydan, Tejaswini Mishra, Jad Nasrini, Brian D Piening, Lindsay F Rizzardi, Kumar Sharma, Jamila H Siamwala, Lynn Taylor, Martha Hotz Vitaterna, Maryam Afkarian, Ebrahim Afshinnekoo, Sara Ahadi, Aditya Ambati, Maneesh Arya, Daniela Bezdan, Colin M Callahan, Songjie Chen, Augustine M K Choi, George E Chlipala, Kévin Contrepois, Marisa Covington, Brian E Crucian, Immaculata De Vivo, David F Dinges, Douglas J Ebert, Jason I Feinberg, Jorge A Gandara, Kerry A George, John Goutsias, George S Grills, Alan R Hargens, Martina Heer, Ryan P Hillary, Andrew N Hoofnagle, Vivian Y H Hook, Garrett Jenkinson, Peng Jiang, Ali Keshavarzian, Steven S Laurie, Brittany Lee-McMullen, Sarah B Lumpkins, Matthew MacKay, Mark G Maienschein-Cline, Ari M Melnick, Tyler M Moore, Kiichi Nakahira, Hemal H Patel, Robert Pietrzyk, Varsha Rao, Rintaro Saito, Denis N Salins, Jan M Schilling, Dorothy D Sears, Caroline K Sheridan, Michael B Stenger, Rakel Tryggvadottir, Alexander E Urban, Tomas Vaisar, Benjamin Van Espen, Jing Zhang, Michael G Ziegler, Sara R Zwart, John B Charles, Craig E Kundrot, Graham B I Scott, Susan M Bailey, Mathias Basner, Andrew P Feinberg, Stuart M C Lee, Christopher E Mason, Emmanuel Mignot, Brinda K Rana, Scott M Smith, Michael P Snyder, Fred W Turek
ABSTRAKT
To understand the health impact of long-duration spaceflight, one identical twin astronaut was monitored before, during, and after a 1-year mission onboard the International Space Station; his twin served as a genetically matched ground control. Longitudinal assessments identified spaceflight-specific changes, including decreased body mass, telomere elongation, genome instability, carotid artery distension and increased intima-media thickness, altered ocular structure, transcriptional and metabolic changes, DNA methylation changes in immune and oxidative stress-related pathways, gastrointestinal microbiota alterations, and some cognitive decline postflight. Although average telomere length, global gene expression, and microbiome changes returned to near preflight levels within 6 months after return to Earth, increased numbers of short telomeres were observed and expression of some genes was still disrupted. These multiomic, molecular, physiological, and behavioral datasets provide a valuable roadmap of the putative health risks for future human spaceflight.