Merck
YAP, but Not RSPO-LGR4/5, Signaling in Biliary Epithelial Cells Promotes a Ductular Reaction in Response to Liver Injury.
Cell stem cell (2019-05-14)
Lara Planas-Paz, Tianliang Sun, Monika Pikiolek, Nadire R Cochran, Sebastian Bergling, Vanessa Orsini, Zinger Yang, Frederic Sigoillot, Jasna Jetzer, Maryam Syed, Marilisa Neri, Sven Schuierer, Lapo Morelli, Philipp S Hoppe, Wibke Schwarzer, Carlos M Cobos, John L Alford, Le Zhang, Rachel Cuttat, Annick Waldt, Nicole Carballido-Perrig, Florian Nigsch, Bernd Kinzel, Thomas B Nicholson, Yi Yang, Xiaohong Mao, Luigi M Terracciano, Carsten Russ, John S Reece-Hoyes, Caroline Gubser Keller, Andreas W Sailer, Tewis Bouwmeester, Linda E Greenbaum, Jesse J Lugus, Feng Cong, Gregory McAllister, Gregory R Hoffman, Guglielmo Roma, Jan S Tchorz
ABSTRAKT
Biliary epithelial cells (BECs) form bile ducts in the liver and are facultative liver stem cells that establish a ductular reaction (DR) to support liver regeneration following injury. Liver damage induces periportal LGR5+ putative liver stem cells that can form BEC-like organoids, suggesting that RSPO-LGR4/5-mediated WNT/β-catenin activity is important for a DR. We addressed the roles of this and other signaling pathways in a DR by performing a focused CRISPR-based loss-of-function screen in BEC-like organoids, followed by in vivo validation and single-cell RNA sequencing. We found that BECs lack and do not require LGR4/5-mediated WNT/β-catenin signaling during a DR, whereas YAP and mTORC1 signaling are required for this process. Upregulation of AXIN2 and LGR5 is required in hepatocytes to enable their regenerative capacity in response to injury. Together, these data highlight heterogeneity within the BEC pool, delineate signaling pathways involved in a DR, and clarify the identity and roles of injury-induced periportal LGR5+ cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Albumin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)