Merck
Quantification Challenges for Atom Probe Tomography of Hydrogen and Deuterium in Zircaloy-4.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada (2019-03-12)
Isabelle Mouton, Andrew J Breen, Siyang Wang, Yanhong Chang, Agnieszka Szczepaniak, Paraskevas Kontis, Leigh T Stephenson, Dierk Raabe, M Herbig, T Ben Britton, Baptiste Gault
ABSTRAKT
Analysis and understanding of the role of hydrogen in metals is a significant challenge for the future of materials science, and this is a clear objective of recent work in the atom probe tomography (APT) community. Isotopic marking by deuteration has often been proposed as the preferred route to enable quantification of hydrogen by APT. Zircaloy-4 was charged electrochemically with hydrogen and deuterium under the same conditions to form large hydrides and deuterides. Our results from a Zr hydride and a Zr deuteride highlight the challenges associated with accurate quantification of hydrogen and deuterium, in particular associated with the overlap of peaks at a low mass-to-charge ratio and of hydrogen/deuterium containing molecular ions. We discuss possible ways to ensure that appropriate information is extracted from APT analysis of hydrogen in zirconium alloy systems that are important for nuclear power applications.