Merck
  • Sulfonate-Functionalized Mesoporous Silica Nanoparticles as Carriers for Controlled Herbicide Diquat Dibromide Release through Electrostatic Interaction.
Sulfonate-Functionalized Mesoporous Silica Nanoparticles as Carriers for Controlled Herbicide Diquat Dibromide Release through Electrostatic Interaction.
International journal of molecular sciences (2019-03-20)
Yongpan Shan, Lidong Cao, Chunli Xu, Pengyue Zhao, Chong Cao, Fengmin Li, Bo Xu, Qiliang Huang
ABSTRAKT
Environmental stimuli-responsive pesticide release is desirable for enhanced efficiency and reduced side effects. In most cases, the loading and release of pesticides mainly depends on hydrophobic interactions and hydrogen bonding. Electrostatic interaction is less investigated as a weapon for achieving high loading content and controlled pesticide release. In this work, negative-charge decorated mesoporous silica nanoparticles (MSNs) were facilely fabricated by introducing sulfonate groups onto MSNs through a post-grafting method. Sulfonate-functionalized MSNs (MSN-SO₃) were synthesized by conversion of epoxy group into sulfonate group using a bisulfite ion as a ring opening reagent. Diquat dibromide (DQ), one of the globally used quaternary ammonium herbicides, was efficiently loaded into these negatively charged MSN-SO₃ nanoparticles. The loading content was increased to 12.73% compared to those using bare MSNs as carriers (5.31%). The release of DQ from DQ@MSN-SO₃ nanoparticles was pH and ionic strength responsive, which was chiefly governed by the electrostatic interactions. Moreover, DQ@MSN-SO₃ nanoparticles exhibited good herbicidal activity for the control of Datura stramonium L., and the bioactivity was affected by the ionic strength of the release medium. The strategy of cargo loading and release dependent on the electrostatic interactions could be generally used for charge-carrying pesticides using carriers possessing opposite charges to mitigate the potential negative impacts on the environment.