Merck
Constitutively Bound EGFR-Mediated Tyrosine Phosphorylation of TLR9 Is Required for Its Ability To Signal.
Journal of immunology (Baltimore, Md. : 1950) (2018-03-14)
Manoj Veleeparambil, Darshana Poddar, Samar Abdulkhalek, Patricia M Kessler, Michifumi Yamashita, Saurabh Chattopadhyay, Ganes C Sen
ABSTRAKT
Mammalian TLRs recognize microbial infection or cell death-associated danger signals and trigger the appropriate cellular response. These responses determine the strength and the outcome of the host-microbe interaction. TLRs are transmembrane proteins located on the plasma or the endosomal membrane. Their ectodomains recognize specific microbial or endogenous ligands, and the cytoplasmic domains interact with specific proteins to activate intracellular signaling pathways. TLR9, an endosomal TLR, is activated by endocytosed DNA. Activated TLR9 recruits the cytoplasmic adapter MyD88 and other signaling proteins to induce the synthesis of inflammatory cytokines and IFN. Uncontrolled activation of TLR9 leads to the undesired overproduction of inflammatory cytokines and consequent pathogenesis. Therefore, appropriate activation and the regulation of TLR9 signaling are critical. Tyrosine (Tyr) phosphorylation of TLR9 is essential for its activation; however, the role of specific Tyr kinases is not clear. In this article, we report that epidermal growth factor receptor (EGFR), a membrane-bound protein Tyr kinase, is essential for TLR9 signaling. Genetic ablation of EGFR or pharmacological inhibition of its kinase activity attenuates TLR9-mediated induction of genes in myeloid and nonmyeloid cell types. EGFR is constitutively bound to TLR9; upon ligand stimulation, it mediates TLR9 Tyr phosphorylation, which leads to the recruitment of MyD88, activation of the signaling kinases and transcription factors, and gene induction. In mice, TLR9-mediated liver injury and death are blocked by an EGFR inhibitor or deletion of the EGFR gene from myeloid cells, which are the major producers of inflammatory cytokines.