Merck
  • Optimization of sample preparation for intact cell mass spectrometry (matrix-assisted laser desorption/ionization linear time-of-flight mass spectrometry) of endophytic Xylaria.
Optimization of sample preparation for intact cell mass spectrometry (matrix-assisted laser desorption/ionization linear time-of-flight mass spectrometry) of endophytic Xylaria.
Rapid communications in mass spectrometry : RCM (2018-03-03)
Luciana da Silva Amaral, Edson Rodrigues-Filho, Christian P Kubicek, Christoph Herwig, Martina Marchetti-Deschmann, Günter Allmaier
ABSTRAKT
Although the fruiting-body of the fungi of the genus Xylaria shows a great variety of morphological characteristics, their mycelial forms are always very similar, imposing difficulties for their identification. Intact cell mass spectrometry (ICMS) using matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS) can be a fast and reliable strategy to support the differentiation/identification of Xylaria species in those cases where fruit-bodies are not available. Many experimental parameters such as sample preparation and culture media are crucial for filamentous fungi analysis by MALDI-TOFMS. For the purposes of this study, we used four matrices (CHCA, DHB, FA and SA) with five different concentrations (0.1, 0.3, 0.5, 1.0 and 2.5%) of TFA in the matrix, the influence of six different culture media (solid and liquid), and three mycelium peptide/protein extraction protocols (acid, basic and thymol-supported solution) to optimize the sample preparation of the endophytic fungus X. arbuscula. It was observed that sinapinic acid (30 mg/mL) dissolved in acetonitrile/0.1% TFA and PDA were the best matrix solution and culture medium, respectively, for the ICMS of X. arbuscula. The formic acid and ammonium bicarbonate (AB) protocols provided similar mass spectra; however, a higher number of peaks were observed using AB extraction. Mass spectra obtained from different thymol-containing solutions (EtOH/aqueous 0.1% TFA and ACN/aqueous 0.1% TFA) show increasing peak abundances at m/z 3000-6500. X. arbuscula could be analyzed by ICMS. However, an extraction step was required to provide suitable MALDI mass spectra. Formic acid-, AB- and thymol-containing solutions were demonstrated to be good cocktails for the extraction of peptide/protein biomarkers from these fungi.