Merck
Evidence for the Formation of Difluoroacetic Acid in Chlorofluorocarbon-Contaminated Ground Water.
Molecules (Basel, Switzerland) (2019-03-17)
Ute Dorgerloh, Roland Becker, Melanie Kaiser
ABSTRAKT
The concentrations of difluoroacetic acid (DFA) and trifluoroacetic acid (TFA) in rainwater and surface water from Berlin, Germany resembled those reported for similar urban areas, and the TFA/DFA ratio in rainwater of 10:1 was in accordance with the literature. In contrast, nearby ground water historically contaminated with 1,1,2-trichloro-1,2,2-trifluoroethane (R113) displayed a TFA/DFA ratio of 1:3. This observation is discussed versus the inventory of microbial degradation products present in this ground water along with the parent R113 itself. A microbial transformation of chlorotrifluoroethylene (R1113) to DFA so far has not been reported for environmental media, and is suggested based on well-established mammalian metabolic pathways.