Merck
  • [18 F]FPyZIDE: A versatile prosthetic reagent for the fluorine-18 radiolabeling of biologics via copper-catalyzed or strain-promoted alkyne-azide cycloadditions.
[18 F]FPyZIDE: A versatile prosthetic reagent for the fluorine-18 radiolabeling of biologics via copper-catalyzed or strain-promoted alkyne-azide cycloadditions.
Journal of labelled compounds & radiopharmaceuticals (2018-12-18)
Mélanie Roche, Simon Specklin, Mylène Richard, Françoise Hinnen, Kevin Génermont, Bertrand Kuhnast
ABSTRAKT
Methods for the radiolabeling of biologics with fluorine-18 have been of interest for several decades. A common approach consists in the preparation of a prosthetic reagent, a small molecule bearing a fluorine-18 that is conjugated with the macromolecule to an appropriate function. Click chemistry, and more particularly cycloadditions, is an interesting approach to radiolabel molecules thanks to mild reaction conditions, high yields, low by-products formation, and strong orthogonality. Moreover, the chemical functions involved in the cycloaddition reaction are stable in the drastic radiofluorination conditions, thus allowing a simple radiosynthetic route to prepare the prosthetic reagent. We report herein the radiosynthesis of 18 F-FPyZIDE, a pyridine-based azide-bearing prosthetic reagent. We exemplified its conjugation via copper-catalyzed cycloaddition (CuAAC) and strain-promoted cycloaddition (SPAAC) with several terminal alkyne or strained alkyne model compounds.