Merck
Does salinity modulates the response of Mytilus galloprovincialis exposed to triclosan and diclofenac?
Environmental pollution (Barking, Essex : 1987) (2019-05-24)
Rosa Freitas, Francesca Coppola, Silvana Costa, Chiara Manzini, Luigi Intorre, Valentina Meucci, Amadeu M V M Soares, Carlo Pretti, Montserrat Solé
ABSTRAKT
In the present study Mytilus galloprovincialis mussels were exposed for 28 days to three salinities: 30 (control), 25 and 35. Simultaneously, organisms at each salinity were exposed to either the antimicrobial agent Triclosan (TCS) or the pharmaceutical drug Diclofenac (DIC) at 1 μg/L. Salinity alone and exposure to PPCPs changed mussel's metabolic capacity and oxidative status, but no additive or synergetic effects resulting from the combined exposures were observed. Overall, the metabolic capacity of mussels was decreased when exposed to TCS and DIC under control salinity, which was less pronounced at salinities out of the control level. TCS had a notorious effect over glutathione peroxidase activity while DIC exposure enhanced catalase response. Such defence mechanisms were able to prevent cellular damage but still a clear reduction in GSH/GSSG ratio after PPCPs exposures indicates oxidative stress which could compromise bivalve's performance to further stressing events.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diclofenac sodium salt