Merck
Elevated levels of soluble PD-L1 are associated with reduced recurrence in papillary thyroid cancer.
Endocrine connections (2019-06-30)
Marra Aghajani, Tara Roberts, Tao Yang, Charles McCafferty, Nicole Caixeiro, Paul DeSouza, Navin Niles
ABSTRAKT
To date, no research evaluating the predictive capabilities of soluble programmed cell death-ligand 1 (sPD-L1) in thyroid cancer patients has been performed. We aimed to investigate the prognostic significance of sPD-L1 expression in papillary thyroid cancer (PTC), and to evaluate the association between sPD-L1 levels with tumoural PD-L1 expression and patient outcomes. Pre-treatment levels of serum and plasma sPD-L1 were measured by enzyme-linked immunosorbent assay (ELISA) in 101 PTC patients. Tissue microarrays were stained with an anti-PD-L1 antibody, clone SP263 (Ventana). The median serum sPD-L1 concentration in PTC patients was significantly higher compared to healthy controls (p=0.028). An increased incidence of extrathyroidal extension was significantly associated with an elevated serum sPD-L1 level (p=0.015). Patients with high serum sPD-L1 levels had significantly shorter median disease-free survival (DFS) as compared to those with low sPD-L1 levels (p=0.011). Following multivariate analysis, serum sPD-L1 was the only statistically significant predictor for DFS. Patients with both positive serum and tumoural PD-L1 expression had a significantly shorter DFS than those in any other subgroup (p=0.007). Our study is the first to confirm that sPD-L1 concentration is significantly associated with patient outcome in PTC. Soluble PD-L1 may provide clinicians with a non-invasive biomarker that can lessen dependence on tissue biopsies and diagnose aggressive thyroid cancers at a more treatable stage.