Merck
  • Rose Bengal incorporated to α-cyclodextrin microparticles for photodynamic therapy against the cariogenic microorganism Streptococcus mutans.
Rose Bengal incorporated to α-cyclodextrin microparticles for photodynamic therapy against the cariogenic microorganism Streptococcus mutans.
Photodiagnosis and photodynamic therapy (2018-11-24)
F J R Alexandrino, E M Bezerra, R F Da Costa, L R L Cavalcante, F A M Sales, T S Francisco, L K A Rodrigues, D H Almeida de Brito, N M P S Ricardo, S N Costa, P de Lima-Neto, I L Barroso-Neto, E W S Caetano, V N Freire
ABSTRAKT
Rose Bengal@α-cyclodextrin (RB@α-CD) microparticles (μPs) were prepared and the RB inclusion in α-CD was experimentally demonstrated through infrared, UV-VIS absorption spectroscopy and cyclic voltammetry. The RB inclusion in α-CD was theoretically investigated using classical molecular mechanics calculations, the simulation results showing that RB can be included in both the narrow and wide apertures of the α-cyclodextrin ring with configurations exhibiting average binding energies of about 27 kcal mol-1. The prepared RB@α-CD microparticles were characterized through Scanning Electron Microscopy (SEM) and it was demonstrated that they are highly efficient in the photodynamic therapy against a Streptococcus mutans (the main bacteria of cariogenic dental plaque) suspension, as a concentration of RB@α-CD μPs 10 times smaller than the usual concentration of pure RB is still capable to produce significant antibacterial activity.