Merck
Structural determinants conferring unusual long life in human serum to rattlesnake-derived antimicrobial peptide Ctn[15-34].
Journal of peptide science : an official publication of the European Peptide Society (2019-07-19)
Clara Pérez-Peinado, Susana A Dias, Diogo A Mendonça, Miguel A R B Castanho, Ana S Veiga, David Andreu
ABSTRAKT
Ctn[15-34], a downsized version of the snake venom cathelicidin-like peptide crotalicidin (Ctn), shows an unusually high lifespan (t1/2 , approximately 12 h) in human serum, which significantly adds to its promise as an antimicrobial and antitumor agent. Herein we investigate the role of Ctn[15-34] structure on serum survival. Using a set of analogs, we show that C-terminal amidation, as well as the specific layout of the Ctn[15-34] sequence-a helical N-terminal domain followed by a hydrophobic domain-is crucial for slow degradation, and any change in their arrangement results in significantly lower t1/2 . Aside from the privileged primary structure, features such as self-aggregation can be ruled out as causes for the long serum life. Instead, studies in other protease-rich fluids suggest a key role for certain human serum components. Finally, we demonstrate that Ctn[15-34] is able to induce bacterial death even after 12-hour pre-incubation in serum, in agreement with the proteolytic data. Altogether, the results shed light on the uncommon stability of Ctn[15-34] in human serum and confirm its potential as an anti-infective lead.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered