- A new menthane-type monoterpenoid from fermented Illigera aromatica with Clonostachys rogersoniana 828H2.
A new menthane-type monoterpenoid from fermented Illigera aromatica with Clonostachys rogersoniana 828H2.
Journal of Asian natural products research (2018-05-12)
Jian-Wei Dong, Le Cai, Xue-Jiao Li, Ya-Xian Shi, Jia-Peng Wang, Rui-Feng Mei, Zhong-Tao Ding
PMID29749266
ABSTRAKT
A new menthane-type monoterpenoid, illigerate E (1), as well as two known ones, (1R*,3R*,4S*,6R*)-6,8-dihydroxymenthol (2) and cis-4-hydroxy-5-(1-hydroxy-1-methylethyl)-2-methyl-2-cyclohexene-1-one (3), were isolated from fermented Illigera aromatica with Clonostachys rogersoniana 828H2. Their structures were identified by HRESIMS and 1D/2D NMR spectra. Their inhibitory effects of NO production in RAW 264.7 macrophages were estimated.