Merck
In-vitro phagocytosis and intracellular killing of Pasteurella multocida B:2 by phagocytic cells of buffaloes.
Microbial pathogenesis (2019-04-13)
Y Puspitasari, S Annas, M N Adza-Rina, M Zamri-Saad
ABSTRAKT
Pasteurella multocida B:2 is a Gram-negative organism causing haemorrhagic septicaemia (HS) in buffaloes. It causes severe pulmonary infection, leading to infiltration of numerous macrophages and neutrophils. Despite the inflammatory response, buffaloes succumb to HS. This study aims to evaluate the in-vitro efficacy of macrophages and neutrophils of buffalo following exposure to P. multocida B:2. In-vitro infections were done using 107 cfu/ml of P. multocida B:2 for Group 1, Escherichia coli for Group 2 and Mannhaemia haemolytica A:2 for Group 3 cells. The inoculated cell cultures were harvested at 0, 30, 60 and 120 min post-exposure and the phagocytic, killing and cell death rates were determined. Both phagocytosis and killing rates of all bacteria increased over time. Phagocytosis involved between 71% and 73% neutrophils and between 60% and 64% macrophages at 120 min. Killing rate of all bacteria involved between 76% and 79% for neutrophils and between 70% and 74% for macrophages at 120 min. Death rate of neutrophils ranged between 67% in Group 3, and 88% in Group 1 at 120 min, significantly (p < 0.05) higher than Group 3 but insignificant (p > 0.05) than Group 2. Similar pattern was observed for death rate of macrophages. The phagocytosis and killing rates of P. multocida B:2 were similar to other bacterial species used in this study but more neutrophils and macrophages were dead following infection by P. multocida B:2 than M. haemolytica A:2.