Merck
  • Prognostic significance of circulating tumor cells (CTCs) in Egyptian non-metastatic colorectal cancer patients: A comparative study for four different techniques of detection (Flowcytometry, CellSearch, Quantitative Real-time PCR and Cytomorphology).
Prognostic significance of circulating tumor cells (CTCs) in Egyptian non-metastatic colorectal cancer patients: A comparative study for four different techniques of detection (Flowcytometry, CellSearch, Quantitative Real-time PCR and Cytomorphology).
Experimental and molecular pathology (2018-12-24)
Abeer A Bahnassy, Salem E Salem, Marwa Mohanad, Nermeen Z Abulezz, Mona S Abdellateif, Marwa Hussein, Chahd A N Zekri, Abdel-Rahman N Zekri, Nasr M A Allahloubi
ABSTRAKT
We assessed CTCs counts in NMCRC patients using four different techniques. CTCs were detected in 63 NMCRC patients, 40 benign bowel diseases (BBD) and 40 normal controls (NC) using, flow-cytometry (FCM), CellSearch (CS), cytomorphology and quantitative real time (qPCR) for CK19, MUC1, CD44, CD133, ALDH1 expression. Results were correlated to progression free (PFS) and overall (OS). Positive CTCs (≥4 cells /7.5 mL blood) were detected in 50.8% (32/63) NMCRC by FCM and 7.5% (3/40) BBD (p < .001). CTCs were detected in 34/63 (54%) NMCRC, 4/40 (10%) BBD (p < .001) by CS. CK19, MUC1, CD44, CD133 and ALDH1 were expressed in 35 (55.6%), 29 (46.0%), 28 (44.4%), 26 (41.3%) and 25 (41.3%) cases of NMCRC. In BBD 4/40 (10%) cases expressed CK19, MUC1 and CD44, while 2/40 (5%) expressed CD133. Cytomorphology showed the lowest sensitivity (47.6%) and specificity (90%) for CTCs detection. The combined use of FCM or CS with CTCs-mRNA markers improved the sensitivity and specificity to 68.3%, and 95.0%; respectively. Positive CTCs and mRNA markers expression were significantly associated with shorter 5-yr PFS and OS. In multivariate analysis, CTCs mRNA markers were independent prognostic factors for PFS and OS. Enumeration of CTCs by FCM and RNA expression for specific colon cancer markers are comparable to CS regarding sensitivity and specificity. CTCs also represent novel therapeutic targets for NMCRC cases.