Merck
Cardiac expression of neutrophil gelatinase-associated lipocalin in a model of cancer cachexia-induced cardiomyopathy.
ESC heart failure (2018-10-28)
Vincenzo Musolino, Sandra Palus, Celine Latouche, Micaela Gliozzi, Francesca Bosco, Federica Scarano, Saverio Nucera, Cristina Carresi, Miriam Scicchitano, Stephan von Haehling, Frederic Jaisser, Gerd Hasenfuss, Stefan D Anker, Vincenzo Mollace, Jochen Springer
ABSTRAKT
Cachexia is a severe consequence of cancer. Although cancer-induced heart atrophy leads to cardiac dysfunction and heart failure (HF), biomarkers for their diagnosis have not been identified. Neutrophil gelatinase-associated lipocalin (NGAL) is an aldosterone-responsive gene increased in HF. We studied NGAL and its association with aldosterone levels in a model of cancer cachexia-induced cardiomyopathy. Rats were injected with Yoshida 108 AH-130 hepatoma cells to induce tumour. Cachectic rats were treated daily, for 16 days, with placebo or with 5 or 50 mg/kg/day of spironolactone. Cardiac function was analysed by echocardiography at baseline and at Day 11. Weight loss and atrophy of lean body and fat mass of cachectic rats were significantly attenuated by spironolactone. Cardiac dysfunction of tumour-bearing rats was improved by spironolactone. Plasma aldosterone was up-regulated from 337 ± 7 pg/mL in sham animals to 591 ± 31 pg/mL in the cachectic rats (P < 0.001 vs. sham). Treatment with 50 or 5 mg/kg/day of spironolactone reduced plasma aldosterone to 396 ± 22 and 391 ± 25 pg/mL (P < 0.01 vs. placebo). Plasma levels of NGAL were also increased in cachectic rats (1.462 ± 0.3603 μg/mL) than in controls (0.0936 ± 6 μg/mL, P < 0.001). Spironolactone treatment (50 mg/kg/day) significantly reduced cardiac mRNA and protein NGAL levels (P < 0.05 and P < 0.001 vs. placebo, respectively). NGAL mRNA and protein levels were overexpressed in cachectic animal hearts treated with placebo, compared with control (P < 0.05 and P < 0.01 vs. sham). Spironolactone treatment at 50 mg/kg/day reduced significantly cardiac NGAL (P < 0.05 and P < 0.001 vs. placebo). Cancer cachexia induced increased levels of aldosterone and NGAL, contributing to worsening cardiac damage in cancer cachexia-induced cardiomyopathy. Spironolactone treatment may greatly attenuate cardiac dysfunction and lean mass atrophy associated with cancer cachexia.