Merck
TRUSS inhibition protects against high fat diet (HFD)-stimulated brain injury by alleviation of inflammatory response.
Biochemical and biophysical research communications (2019-02-16)
Qing Zhu, Yong-Yi Zhu, Wei-Ning Wang
ABSTRAKT
High fat diet (HFD)-induced obesity is associated with insulin resistance (IR) and other chronic, diet associated illnesses, including neuroinflammation and brain injury. However, the involvement of inflammatory response in HFD-elicited central nerve injury has yet to be fully determined. Recent studies have indicated that tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein (TRUSS), also known as TRPC4AP, plays an essential role in regulating inflammation via the meditation of NF-κB signaling. In the present study, we attempted to explore the effects of TRUSS on HFD-induced brain injury in the wild type mice (TRUSS+/+) or TRUSS-knockout mice (TRUSS-/-). The results suggested that TRUSS deletion attenuated HFD-induced cognitive impairments in mice. HFD-elicited metabolic disorders were also highly improved by the loss of TRUSS, as evidenced by the reduced serum glucose and insulin levels, as well as the lipid deposition in liver tissues. In addition, HFD-triggered brain injury was markedly alleviated by the TRUSS ablation, as proved by the reduction of GFAP and Iba1 expressions in hippocampus and hypothalamus. Moreover, TRUSS-/- mice exhibited a significant decrease in the expression of pro-inflammatory cytokines, accompanied with the inactivation of IKKα/IκBα/NF-κB pathway. At the same time, HFD-induced dyslipidemia was also alleviated by the loss of TRUSS. The in vitro study verified the protective effects of TRUSS-suppression against HFD-induced central nerve injury and hepatic steatosis by restraining the inflammatory response. In summary, our data indicated that TRUSS participated in metabolic syndrome-induced brain injury and pointed to the repression of TRUSS as a promising strategy for cognitive deficits therapy.