Merck
Hyaluronan synthase 2-mediated hyaluronan production mediates Notch1 activation and liver fibrosis.
Science translational medicine (2019-06-14)
Yoon Mee Yang, Mazen Noureddin, Cheng Liu, Koichiro Ohashi, So Yeon Kim, Divya Ramnath, Elizabeth E Powell, Matthew J Sweet, Yoon Seok Roh, I-Fang Hsin, Nan Deng, Zhenqiu Liu, Jiurong Liang, Edward Mena, Daniel Shouhed, Robert F Schwabe, Dianhua Jiang, Shelly C Lu, Paul W Noble, Ekihiro Seki
ABSTRAKT
Hyaluronan (HA), a major extracellular matrix glycosaminoglycan, is a biomarker for cirrhosis. However, little is known about the regulatory and downstream mechanisms of HA overproduction in liver fibrosis. Hepatic HA and HA synthase 2 (HAS2) expression was elevated in both human and murine liver fibrosis. HA production and liver fibrosis were reduced in mice lacking HAS2 in hepatic stellate cells (HSCs), whereas mice overexpressing HAS2 had exacerbated liver fibrosis. HAS2 was transcriptionally up-regulated by transforming growth factor-β through Wilms tumor 1 to promote fibrogenic, proliferative, and invasive properties of HSCs via CD44, Toll-like receptor 4 (TLR4), and newly identified downstream effector Notch1. Inhibition of HA synthesis by 4-methylumbelliferone reduced HSC activation and liver fibrosis in mice. Our study provides evidence that HAS2 actively synthesizes HA in HSCs and that it promotes HSC activation and liver fibrosis through Notch1. Targeted HA inhibition may have potential to be an effective therapy for liver fibrosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%