Merck
Simulating Transplant Small-for-size Grafts Using Human Liver Monosegments: The Impact of Portal Perfusion Pressure.
Transplantation proceedings (2019-02-10)
M Mohamed, L Kang, C Zhang, B Edenfield, J Sykes, T Brown, J L Johnson, F Rehman, J H Nguyen
ABSTRAKT
Small-for-size-liver grafts (SFSG) in adult transplant recipients have elevated risk of graft failure, limiting its application in clinical liver transplantation. Relevant preclinical model of SFSG is lacking. Relevant to deceased-donor split liver transplant and living-donor liver transplant in adult recipients, in this study, we present our initial characterization of SFSG model using monosegments of a discarded human donor liver.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%