Merck
Production of santalenes and bergamotene in Nicotiana tabacum plants.
PloS one (2019-01-05)
Jun-Lin Yin, Woon-Seng Wong
ABSTRAKT
Terpenes play an important role in plant-insect relationships, and these relationships can potentially be modified by altering the profile of terpenes emitted from plants using metabolic engineering methods. Transgenic plants generated by employing such methods offer the prospect of low-cost sustainable pest management; in this regard, we used chloroplast targeting and cytosolic mevalonic acid pathway enhancement in this study to investigate the interaction of santalenes and bergamotene with insects. The santalene- and bergamotene-emitting transgenic tobacco plants thus generated were utilized to study host preference in the green peach aphid (Myzus persicae (Sulzer)). The results showed that co-expression of either 3-hydroxy-3-methylglutaryl-CoA reductase (HMGR) or truncated HMGR with santalene synthase led to the production of higher amounts of santalenes and bergamotene in transgenic tobacco plants, and that these santalene- and bergamotene-emitting plants were attractive to green peach aphids. We accordingly propose that such transgenic plants may have potential application in pest management as a trap crop to prevent green peach aphid infestation of wild-type tobacco plants.