Merck
Synthesis of imprinted hydrogel microbeads by inverse Pickering emulsion to controlled release of adenosine 5'‑monophosphate.
Materials science & engineering. C, Materials for biological applications (2019-04-29)
Mohamed G Ayari, Porkodi Kadhirvel, Patrick Favetta, Bernard Plano, Corinne Dejous, Benjamin Carbonnier, Luigi A Agrofoglio
ABSTRAKT
Herein, we propose the synthesis of a microspherical imprinted hydrogel meant for the controlled release of a nucleotide, adenosine 5'-monophosphate (5'-AMP). Indeed, molecularly imprinted polymers-based (MIPs) materials possess remarkable selective molecular recognition ability that mimicks biological systems. MIPs have been used in numerous applications and hold great promise for the vectorization and/or controlled release of therapeutics and cosmetics. But, the conception of imprinted hydrogels-based drug delivery systems that are able to release polar bioactive compounds is explored weakly. Herein, the synthesis of imprinted hydrogel microbeads by inverse Pickering emulsion is detailed. Microspheres showed a large 5'-AMP loading capacity, around 300 mg·g-1, and a high binding capacity comparatively to the non-imprinted counterpart. The MIP had a thermo-responsive release behavior providing sustained release of adenosine 5'-monophosphate in an aqueous buffer simulating both human skin pH and temperature.