Merck
Corticosteroid signaling at the brain-immune interface impedes coping with severe psychological stress.
Science advances (2019-06-01)
A Kertser, K Baruch, A Deczkowska, A Weiner, T Croese, M Kenigsbuch, I Cooper, M Tsoory, S Ben-Hamo, I Amit, M Schwartz
ABSTRAKT
The immune system supports brain plasticity and homeostasis, yet it is prone to changes following psychological stress. Thus, it remains unclear whether and how stress-induced immune alterations contribute to the development of mental pathologies. Here, we show that following severe stress in mice, leukocyte trafficking through the choroid plexus (CP), a compartment that mediates physiological immune-brain communication, is impaired. Blocking glucocorticoid receptor signaling, either systemically or locally through its genetic knockdown at the CP, facilitated the recruitment of Gata3- and Foxp3-expressing T cells to the brain and attenuated post-traumatic behavioral deficits. These findings functionally link post-traumatic stress behavior with elevated stress-related corticosteroid signaling at the brain-immune interface and suggest a novel therapeutic target to attenuate the consequences of severe psychological stress.