Merck
Cyhalothrin biodegradation in Cunninghamella elegans.
Environmental science and pollution research international (2018-11-15)
William Palmer-Brown, Paula Letícia de Melo Souza, Cormac D Murphy
ABSTRAKT
The insecticide λ-cyhalothrin was incubated with planktonic and biofilm cultures of the fungus Cunninghamella elegans. 19F nuclear magnetic resonance spectroscopy demonstrated that the compound was initially biosorbed to the biomass and more slowly degraded by the fungus. Furthermore, the presence of trifluoromethyl-containing metabolites was observed. Analysis of culture extracts by gas chromatography-mass spectrometry (GC-MS) identified non-fluorinated metabolites that suggested the likely catabolic pathway. The hydroxylated metabolites were probably generated from the action of cytochromes P450 (CYPs), as the presence of CYP inhibitors resulted in the absence of biodegradation. Planktonic cells were measurably faster at degrading the pesticide compared with biofilm.