Merck
Abnormalities in cortical interneuron subtypes in ephrin-B mutant mice.
The European journal of neuroscience (2018-06-16)
Asghar Talebian, Rachel Britton, Mark Henkemeyer
ABSTRAKT
To explore roles for ephrin-B/EphB signaling in cortical interneurons, we previously generated ephrin-B (Efnb1/b2/b3) conditional triple mutant (TMlz ) mice using a Dlx1/2.Cre inhibitory neuron driver and green fluorescent protein (GFP) reporters for the two main inhibitory interneuron groups distinguished by expression of either glutamic acid decarboxylase 1 (GAD1; GAD67-GFP) or 2 (GAD2; GAD65-GFP). This work showed a general involvement of ephrin-B in migration and population of interneurons into the embryonic neocortex. We now determined whether specific interneurons are selectively affected in the adult brains of TMlz .Cre mice by immunostaining with antibodies that identify the different subtypes. The results indicate that GAD67-GFP-expressing interneurons that also express parvalbumin (PV), calretinin (CR) and, to a lesser extent, somatostatin (SST) and Reelin (Rln) were significantly reduced in the cortex and hippocampal CA1 region in TMlz .Cre mutant mice. Neuropeptide Y (NPY) interneurons that also express GAD67-GFP were reduced in the hippocampal CA1 region, but much less so in the cortex, although these cells exhibited abnormal cortical layering. In GAD65-GFP-expressing interneurons, CR subtypes were reduced in both cortex and hippocampal CA1 region, whereas Rln interneurons were reduced exclusively in hippocampus, and the numbers of NPY and vasoactive intestinal polypeptide (VIP) subtypes appeared normal. PV and CR subtype interneurons in TMlz .Cre mice also exhibited reductions in their perisomatic area, suggesting abnormalities in dendritic/axonal complexity. Altogether, our data indicate that ephrin-B expression within forebrain interneurons is required in specific subtypes for their normal population, cortical layering and elaboration of cell processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse