Merck
  • Effect of photoperiodic alterations on depression-like behavior and the brain serotonin system in mice genetically different in tryptophan hydroxylase 2 activity.
Effect of photoperiodic alterations on depression-like behavior and the brain serotonin system in mice genetically different in tryptophan hydroxylase 2 activity.
Neuroscience letters (2019-01-28)
Ekaterina Y Bazhenova, Dariya V Fursenko, Elizabeth A Kulikova, Nikita V Khotskin, Nadezhda A Sinyakova, Alexander A Kulikov
ABSTRAKT
Reduction of natural illumination in fall/winter months causes seasonal affective disorders (SAD) in vulnerable individuals. Neurotransmitter serotonin (5-HT) is involved in the mechanism of SAD. Tryptophan hydroxylase-2 (TPH2) is the key enzyme of 5-HT synthesis in the brain. C1473 G polymorphism in the Tph2 gene is a key factor defining the enzyme activity in the mouse brain. The main aims of the study were to investigate the effects of C1473 G polymorphism on behavior and brain 5-HT system responses to photoperiod alterations. The experiment was carried out on adult mouse males of B6-1473C and B6-1473 G congenic lines with normal and low TPH2 activities, respectively. B6-1473C and B6-1473 G mice were divided into four groups of 8 each and exposed for 28 days to standard-day (14 h light and 10 h darkness) or short-day (4 h light and 20 h darkness) conditions. No effect of photoperiod on locomotor, exploratory activities and anxiety in the open field test was observed. At the same time, photoperiod alterations affected depressive-like immobility in the forced swim test, the 5-HT, 5-hydroxyindoleacetic acid (5-HIAA) levels, 5-HIAA/5-HT ratio and the Htr2a mRNA level in hippocampus and midbrain. The effect of the interaction between C1473 G polymorphism and photoperiod on 5-HT level and 5-HIAA/5-HT ratio in hippocampus was revealed. Short-day conditions reduced the level and increased 5-HIAA/5-HT ratio in this structure only in B6-1473 G mice. At the same time, C1473 G polymorphism does not alter effects of short-day conditions on immobility time in the forced swim test and the Htr2a mRNA level in the brain.