Merck
  • Mesenchymal Stromal Cells Derived from Normal Cervix and Cervical Cancer Tumors Increase CD73 Expression in Cervical Cancer Cells Through TGF-β1 Production.
Mesenchymal Stromal Cells Derived from Normal Cervix and Cervical Cancer Tumors Increase CD73 Expression in Cervical Cancer Cells Through TGF-β1 Production.
Stem cells and development (2019-01-31)
Luis Roberto Ávila-Ibarra, María de Lourdes Mora-García, Rosario García-Rocha, Jorge Hernández-Montes, Benny Weiss-Steider, Juan José Montesinos, Marcela Lizano Soberon, Patricia García-López, Christian Azucena Don López, Daniela Berenice Torres-Pineda, Rommel Chacón-Salinas, Luis Vallejo-Castillo, Sonia Mayra Pérez-Tapia, Alberto Monroy-García
ABSTRAKT
Mesenchymal stromal cells (MSCs) in the tumor microenvironment (TME) participate together with tumor cells to suppress antitumor effector cells through the production of immunosuppressive factors, such as transforming growth factor-beta 1 (TGF-β1). Furthermore, TGF-β1 can induce 5'-nucleotidase (CD73) expression in various cell types; this functional activity is associated with the production of adenosine (Ado), which is an immunosuppressive nucleoside. In this study, we provide evidence that coculture of MSCs derived from cervical tumors (CeCa-MSC) with CeCa tumor cells increases CD73 expression in tumor cells and the capacity of these cells to generate Ado in a MSC ratio-dependent manner. Interestingly, the increase in CD73 in the CeCa cell membrane corresponded to an increase in the TGF-β1 expression level in the tumor cells and the TGF-β1 content in the supernatants of the CeCa/CeCa-MSC cocultures. The addition of anti-hTGF-β neutralizing antibodies strongly reversed CD73 expression in the tumor cells. This phenomenon was not exclusive to CeCa-MSCs; coculture of MSCs derived from the normal cervix with CeCa cells produced similar results. These results suggest that the interaction of MSCs with CeCa tumor cells in the TME may condition higher TGF-β1 production to maintain an immunosuppressive status not only through the activity of this cytokine per se but also through its ability to induce CD73 expression in tumor cells and generate an immunosuppressive microenvironment rich in Ado.