Merck
Rho-of-plant activated root hair formation requires Arabidopsis YIP4a/b gene function.
Development (Cambridge, England) (2019-02-17)
Delphine Gendre, Anirban Baral, Xie Dang, Nicolas Esnay, Yohann Boutté, Thomas Stanislas, Thomas Vain, Stéphane Claverol, Anna Gustavsson, Deshu Lin, Markus Grebe, Rishikesh P Bhalerao
ABSTRAKT
Root hairs are protrusions from root epidermal cells with crucial roles in plant soil interactions. Although much is known about patterning, polarity and tip growth of root hairs, contributions of membrane trafficking to hair initiation remain poorly understood. Here, we demonstrate that the trans-Golgi network-localized YPT-INTERACTING PROTEIN 4a and YPT-INTERACTING PROTEIN 4b (YIP4a/b) contribute to activation and plasma membrane accumulation of Rho-of-plant (ROP) small GTPases during hair initiation, identifying YIP4a/b as central trafficking components in ROP-dependent root hair formation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2