Merck
Potential diagnostic value of a type X collagen neo-epitope biomarker for knee osteoarthritis.
Osteoarthritis and cartilage (2019-01-18)
Y He, T Manon-Jensen, L Arendt-Nielsen, K K Petersen, T Christiansen, J Samuels, S Abramson, M A Karsdal, M Attur, A C Bay-Jensen
ABSTRAKT
Phenotypic changes of chondrocytes toward hypertrophy might be fundamental in the pathogenesis of osteoarthritis (OA), of which type X collagen (Col10) is a well-known marker. The purpose was to develop a specific immunoassay for blood quantification of a newly identified neo-epitope of type Col10 to assess its diagnostic value for radiographic knee OA. A neo-epitope of Col10 was identified in urine samples from OA patients. A monoclonal antibody against the neo-epitope was produced in Balb/C mice. The enzyme responsible for the cleavage was identified. Immunohistochemical detection of this neo-epitope was performed on human OA cartilage. An immunoassay (Col10neo) was developed and quantified in two clinical studies: the C4Pain-003 and the NYU OA progression study. Receiver operating characteristic curve (ROC) curve analysis was carried out to evaluate the discriminative power of Col10neo between OA and rheumatoid arthritis (RA). A neo-epitope specific mAb was produced. The Cathepsin K-generated neo-epitope was localized to the pericellular matrix of chondrocytes, while its presence was extended and more prominent in superficial fibrillation in the cartilage with advanced degradation. In the C4Pain study, a higher level of Col10neo was seen in subjects with greater KL grade. The group of the highest tertile of Col10neo included more subjects with KL3-4. In the NYU study, Col10neo was statistically higher in OA than control or RA. ROC curve analysis revealed area under the curve was 0.88 (95% CI 0.81-0.94). Our findings indicate that Col10neo linked to hypertrophic chondrocytes could be used as a diagnostic biochemical marker for knee OA.