Merck
miR-338-3p regulates osteoclastogenesis via targeting IKKβ gene.
In vitro cellular & developmental biology. Animal (2019-03-20)
Dequn Niu, Zheng Gong, Xuemin Sun, Jianchang Yuan, Tiantian Zheng, Xun Wang, Xu Fan, Yingji Mao, Xianfu Liu, Baoding Tang, Yingxiao Fu
ABSTRAKT
This study determined the effects of miR-338-3p on osteoclast (OC) differentiation and activation. The change levels of miR-338-3p in differentiated OCs were investigated by microRNA microarray assay and quantitative real-time PCR analysis. The effects of miR-338-3p on the differentiation and activation of OCs were determined by tartrate-resistant acid phosphatase staining resorption activity assay and Western blot. Target genes of miR-338-3p were identified by target gene prediction and dual-luciferase reporter gene detection assay as well as Western blot. Results showed that miR-338-3p was markedly downregulated in differentiated OCs. miR-338-3p could inhibit the formation and absorption activity of OCs. Western blot showed that miR-338-3p could influence the change levels of OC differentiation-related proteins. Dual-luciferase reporter gene detection assay and Western blot both showed that miR-338-3p directly targeted IKKβ gene. In conclusion, miR-338-3p may affect the formation and activity of OCs by targeting the IKKβ gene.