- A Heparan Sulfate Device for the Regeneration of Osteochondral Defects.
A Heparan Sulfate Device for the Regeneration of Osteochondral Defects.
Tissue engineering. Part A (2018-10-24)
Jonathan H Lee, Xiaoman Luo, Xiafei Ren, Tuan Chun Tan, Raymond A A Smith, Kunchithapadam Swaminathan, Sakthivel Sekar, Kishore Bhakoo, Victor Nurcombe, James H Hui, Simon M Cool
PMID30351222
ABSTRAKT
Repairing damaged joint cartilage remains a significant challenge. Treatment involving microfracture, tissue grafting, or cell therapy provides some benefit, but seldom regenerates lost articular cartilage. Providing a point-of-care solution that is cell and tissue free has the potential to transform orthopedic treatment for such cases. Glycosaminoglycans such as heparan sulfate (HS) are well suited for this purpose because they provide a matrix that enhances the prochondrogenic activities of growth factors normally found at sites of articular damage. In this study, we show the potential of a novel HS device, which is free of exogenous cells or growth factors, in regenerating osteochondral defects.