Merck
Identification of Redox Partners of the Thiol-Disulfide Oxidoreductase SdbA in Streptococcus gordonii.
Journal of bacteriology (2019-02-26)
Naif A Jalal, Lauren Davey, Scott A Halperin, Song F Lee
ABSTRAKT
We previously identified a novel thiol-disulfide oxidoreductase, SdbA, in Streptococcus gordonii that formed disulfide bonds in substrate proteins and played a role in multiple phenotypes. In this study, we used mutational, phenotypic, and biochemical approaches to identify and characterize the redox partners of SdbA. Unexpectedly, the results showed that SdbA has multiple redox partners, forming a complex oxidative protein-folding pathway. The primary redox partners of SdbA that maintain its active site in an oxidized state are a surface-exposed thioredoxin family lipoprotein called SdbB (Sgo_1171) and an integral membrane protein annotated as CcdA2. Inactivation of sdbB and ccdA2 simultaneously, but not individually, recapitulated the sdbA mutant phenotype. The sdbB-ccdA2 mutant had defects in a range of cellular processes, including autolysis, bacteriocin production, genetic competence, and extracellular DNA (eDNA) release. AtlS, the natural substrate of SdbA produced by the sdbB-ccdA2 mutant lacked activity and an intramolecular disulfide bond. The redox state of SdbA in the sdbB-ccdA2 mutant was found to be in a reduced form and was restored when sdbB and ccdA2 were knocked back into the mutant. In addition, we showed that SdbB formed a disulfide-linked complex with SdbA in the cell. Recombinant SdbB and CcdA2 exhibited oxidase activity and reoxidized reduced SdbA in vitro Collectively, our results demonstrate that S. gordonii uses multiple redox partners for oxidative protein folding.IMPORTANCEStreptococcus gordonii is a commensal bacterium of the human dental plaque. Previously, we identified an enzyme, SdbA, that forms disulfide bonds in substrate proteins and plays a role in a number of cellular processes in S. gordonii Here, we identified the redox partners of SdbA. We showed that SdbA has multiple redox partners, SdbB and CcdA2, forming a complex oxidative protein-folding pathway. This pathway is essential for autolysis, bacteriocin production, genetic competence, and extracellular DNA (eDNA) release in S. gordonii These cellular processes are considered to be important for the success of S. gordonii as a dental plaque organism. This is the first example of an oxidative protein-folding pathway in Gram-positive bacteria that consists of an enzyme that uses multiple redox partners to function.