Merck
Inhibiting a spinal cord signaling pathway protects against ischemia injury in rats.
The Journal of thoracic and cardiovascular surgery (2018-09-10)
Jia Huo, Rui Ma, Xin Chai, Hong-Jin Liang, Peng Jiang, Xiao-Ling Zhu, Xin Chen, Bin-Xiao Su
ABSTRAKT
The aim of the study was to examine whether the cannabinoid agonist WIN55212-2 could attenuate ischemic spinal cord injury (SCI) in rats through inhibition of GAPDH/Siah1 signaling. Male Sprague-Dawley rats were distributed randomly into 5 groups: (1) sham group that received no aortic occlusion and injected intraperitoneally (i.p.) with vehicle control after reperfusion; (2) control group that received a 12-minute aortic occlusion and injected i.p. with vehicle control after reperfusion; (3) WIN55212-2 group (WIN) that received the aortic occlusion and injected i.p. with 1 mg/kg of WIN55212-2 after reperfusion; and (4) WIN55212-2 plus AM251 group and (5) WIN55212-2 plus AM630 group that received the same surgical operation as the WIN group, except that 1 mg/kg of AM251 or AM630 was injected i.p. 30 min before each dose of WIN55212-2 injection, respectively. Neurologic function was assessed 48 hours after reperfusion. Histopathologic examination was performed to determine the number of normal neurons in anterior spinal cord. Protein expression of active caspase-3, total caspase-3, glyceraldehyde 3-phosphate dehydrogenase (GAPDH), inducible nitric oxide synthase (iNOS), nuclear factor kappa light chain enhancer of activated B cells (NF-κB), Siah1, tumor necrosis factor α, and interleukin 1β were determined with Western blot and enzyme-linked immunosorbent assay; coimmunoprecipitation assays were also used to determine GAPDH/Siah1 complexing. Finally, terminal deoxynucleotidyl transferase dUTP nick end labeling staining was used to determine neuronal apoptosis in the lumbar spinal cord. The nuclear translocation of GAPDH and Siah1 in the spinal cord was initiated after ischemic spinal cord injury (SCI) along with the increased formation of GAPDH/Siah1 complexes. However, the activation of GAPDH/Siah1 was blocked by WIN. In addition, the treatment of WIN55212-2 promoted neuronal survival in the spinal cord, reduced apoptosis and inflammation, and improved neurologic scores. Furthermore, these beneficial effects of WIN55212-2 were abolished by the combined treatment of the CB2 antagonist AM630, but not the CB1 antagonist AM251. Our findings reveal GAPDH/Siah1 signaling cascades as a novel therapeutic target for ischemic SCI and identify WIN55212-2 with the potential to treat ischemic SCI by targeting this pathway.