Merck
CRISPR/Cas9-mediated knockout of Lcn2 effectively enhanced CDDP-induced apoptosis and reduced cell migration capacity of PC3 cells.
Life sciences (2019-06-21)
Sina Rahimi, Amaneh Mohammadi Roushandeh, Ammar Ebrahimi, Ali Akbar Samadani, Yoshikazu Kuwahara, Mehryar Habibi Roudkenar
ABSTRAKT
Lipocalin 2 (Lcn2/NGAL) belongs to lipocalin superfamily with diverse functions. The precise function of Lcn2, particularly in cancer development, remains to be elucidated yet. In an attempt to knockout of Lcn2 expression by CRISPR/Cas 9 technology in a highly aggressive and invasive prostate cancer cell line and to evaluate the combination therapy with cisplatin (CDDP), this study was conducted. Control CRISPR/Cas9 plasmid and homology-directed repair plasmid or validated human Lcn2 CRISPR/Cas9 KO plasmids were co-transfected into PC3 cells using fugene HD transfection reagent. The stable cells were selected in the presence of puromycin. Correspondingly, knock out of Lcn2 was evaluated by RT-PCR, ELISA, and immunocytochemistry. PC3-Scr (control) and Lcn2-KO (PC3 cells in which lcn2 has been knocked out) were treated with or without cisplatin (CDDP). Cell proliferative ability was measured by WST-1 and colony-formation assays. Apoptosis was evaluated by DAPI staining, in situ cell death detection (TUNEL) assay, and cell death detection ELISA plus methods. The migration capabilities were studied by wound healing/scratch and transwell assays. Lcn2 knock out in a highly aggressive and invasive cancer cell like PC3 decreased cell proliferation and increased the sensitivity of CDDP. Conspicuously, loss of Lcn2 expression effectively enhanced CDDP-induced apoptosis in PC3 cells. Lcn2 knock out by CRISPR/Cas9 technology decreased the cell migration capacity of PC3 cells as well. Lcn2 not only is a valuable and useful biomarker for diagnosis and prognosis of prostate cancer but also and more importantly is a potential novel emerging therapeutic target.