Merck
Effect of the Nano-Ca(OH)2 Addition on the Portland Clinker Cooking Efficiency.
Materials (Basel, Switzerland) (2019-06-05)
Azzurra Zucchini, Paola Comodi, Alessandro Di Michele, Riccardo Vivani, Lucia Mancini, Gabriele Lanzafame, Serena Casagrande, Silvia Gentili, Francesco Vetere, Luca Bartolucci, Gianluca Polidori, Fabio Santinelli, Alessandro Neri
ABSTRAKT
A new technology was tested to improve the cooking efficiency of the raw mixture for Portland clinker production by the use of nano-Ca(OH)2. A decrease in the free lime concentration after the firing of approximately 35% and 55% in the nano-added clinkers burned at 1350 °C and 1450 °C, respectively, with respect to the standard Portland clinkers was observed. Moreover, in the nano-added clinkers, a slight decrease in alite (C3S), of approximately 2-4 wt%, and increase in belite (C2S), of approximately 5-6 wt%, were observed. Despite these variations, the C2S and C3S abundance lies within the ranges for standard Portland clinkers. The results showed that the nano-addition leads to an increase of the raw mixtures' cooking efficiency. The relatively low energy required for the clinker firing could be used to increase the plant productivity and decrease the CO2 emissions during clinker burning. The decrease of the work index of the clinkers produced by the use of the nano-Ca(OH)2 also contributes to the energy saving during clinker grinding. Differences were also found in the pore size distribution among nano-added clinkers and the standard Portland clinker. The smallest porosities with the modal volume lying in the class of 3∙10-6 mm3 were found to increase by the use of nano-Ca(OH)2. However, the pore volumes higher than 2.0∙10-5 mm3 decreased in the nano-added clinkers.