Merck
Photophysical and photodynamic properties of Pyronin Y in micellar media at different temperatures.
Luminescence : the journal of biological and chemical luminescence (2019-03-15)
Burcu Meryem Beşer, Mustafa Arik, Yavuz Onganer
ABSTRAKT
In this study, photophysics and photodynamical properties of Pyronin Y (PyY) in different liquid media were investigated. Interactions of PyY, which is a positively charged pigment compound pertaining to the xanthene derivatives with surfactants possessing distinct charges, were determined by using the molecular absorption and fluorescence spectroscopy techniques. It was observed that band intensities of absorption and fluorescence spectra belonging to PyY increase in proportion to the water when compared to three micelle systems, cetyltrimethylammonium bromide (CTAB), sodium dodecyl sulfate (SDS) and Triton X-100 (TX-100). This suggests that interactions in micelle systems are different from those in deionized water, and solvation and surface interactions modify. It is determined that the strongest interaction occurs between PyY dye and SDS, anionic surfactant, and this interaction arises from the electrostatic forces. Calculated photophysical parameters indicated that the microenvironment of PyY in SDS micelle is different to that of other systems. In temperature studies, it was reported that increasing the temperature of the samples increased non-radiative transitions. Steady-state fluorescence anisotropy values were calculated by using fluorescence intensities of PyY compound in pre-micellar, micellar and post-micellar systems. Once the PyY fluorescence probe is added to the surfactant containing solutions below the critical micelle concentrations, the measured anisotropy values were found to be low because the probe remains in the deionized water phase. When the surfactant concentration of the medium becomes closer to the critical micelle concentrations, the steady-state anisotropy value prominently increases. This is because of the restrictions on the rotational diffusion of the probe in micellar solution. It is observed that positively charged PyY shows a higher affinity to the negatively charged SDS compared with the positively charged CTAB and neutral TX-100 surfactants. This can be explained by Coulombic interactions.