Merck
Anti-fumarase antibody promotes the dropout of photoreceptor inner and outer segments in diabetic macular oedema.
Diabetologia (2018-11-30)
Shin Yoshitake, Tomoaki Murakami, Kiyoshi Suzuma, Tatsuya Yoshitake, Akihito Uji, Satoshi Morooka, Yoko Dodo, Masahiro Fujimoto, Yang Shan, Patrice E Fort, Shinji Ito, Akitaka Tsujikawa, Nagahisa Yoshimura
ABSTRAKT
In diabetic macular oedema (DMO), blood components passing through the disrupted blood-retinal barrier cause neuroinflammation, but the mechanism by which autoantibodies induce neuroglial dysfunction is unknown. The aim of this study was to identify a novel autoantibody and to evaluate its pathological effects on clinically relevant photoreceptor injuries. Biochemical purification and subsequent peptide fingerprinting were applied to identify autoantigens. The titres of autoantibodies in DMO sera were quantified and their associations with clinical variables were evaluated. Two animal models (i.e. passive transfer of autoantibodies and active immunisation) were characterised with respect to autoimmune mechanisms underlying photoreceptor injuries. After screening serum IgG from individuals with DMO, fumarase, a Krebs cycle enzyme expressed in inner segments, was identified as an autoantigen. Serum levels of anti-fumarase IgG in participants with DMO were higher than those in diabetic participants without DMO (p < 0.001) and were related to photoreceptor damage and visual dysfunction. Passively transferred fumarase IgG from DMO sera in concert with complement impaired the function and structure of rodent photoreceptors. This was consistent with complement activation in the damaged photoreceptors of mice immunised with fumarase. Fumarase was recruited to the cell surface by complement and reacted to this autoantibody. Subsequently, combined administration of anti-fumarase antibody and complement elicited mitochondrial disruption and caspase-3 activation. This study has identified anti-fumarase antibody as a serum biomarker and demonstrates that the generation of this autoantibody might be a pathological mechanism of autoimmune photoreceptor injuries in DMO.